Ethyl pyrimidine-4-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-pyrimidinecarboxylate
OR953072-1G
OR953072-25G
OR953072-5G
APOSOR953072-1GEA
56.7
EUR
APOSOR953072-1G
APOSOR953072-25G
APOSOR953072-5G
Ethyl pyrimidine-4-carboxylate 95%
Ethyl pyrimidine-4-carboxylate
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832999 CAS-Nummer: 62846-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...