N,N-Dimethyl-3-nitropyridin-2-amine 96%

Lieferant: Apollo Scientific

OR953155-5G
APOSOR953155-5GEA 89.9 EUR
APOSOR953155-5G
N,N-Dimethyl-3-nitropyridin-2-amine 96%
N,N-Dimethyl-3-nitropyridin-2-amine
Formel: C₇H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00956024
CAS-Nummer: 5028-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR