3-Ethylanilin 95%

Lieferant: Apollo Scientific

OR953182-100G OR953182-25G
APOSOR953182-100GEA 227 EUR
APOSOR953182-100G APOSOR953182-25G
3-Ethylanilin 95%
3-Ethylanilin
Formel: C₈H₁₁N
Molecular Weight: 121,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007818
CAS-Nummer: 587-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR