2,5-Dichlor-α-methylbenzylalkohol 95%

Lieferant: Apollo Scientific

OR953205-25G
APOSOR953205-25GEA 111 EUR
APOSOR953205-25G
2,5-Dichlor-α-methylbenzylalkohol 95%
2,5-Dichlor-α-methylbenzylalkohol
Formel: C₈H₈Cl₂O
Molecular Weight: 191.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021861
CAS-Nummer: 1475-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR