Methyl-2-amino-2-methylpropanoate hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl alpha-aminoisobutyrate hydrochloride, H-Aib-OMe.HCl, Methyl 2-aminoisobutyrate hydrochloride, H-Aib-OMe·HCl
OR953353-100G
OR953353-25G
OR953353-5G
OR953353-500G
APOSOR953353-100GEA
71.2
EUR
APOSOR953353-100G
APOSOR953353-25G
APOSOR953353-5G
APOSOR953353-500G
Methyl-2-amino-2-methylpropanoate hydrochloride 95%
Methyl-2-amino-2-methylpropanoate hydrochloride
Formel:
C₅H₁₁NO₂·HCl Molecular Weight: 153,6 g/mol Schmelzpunkt: 185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00214247 CAS-Nummer: 15028-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...