6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine 98%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-3-methyl-[1,2,4]triazolo[4,3-b]pyridazine
OR953373-1G
OR953373-5G
APOSOR953373-1GEA
150
EUR
APOSOR953373-1G
APOSOR953373-5G
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine 98%
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine
Formel:
C₆H₅ClN₄ Molecular Weight: 168,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975127 CAS-Nummer: 7197-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...