4-Acetyl-1H-pyrrole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR953395-1G OR953395-250MG
APOSOR953395-1GEA 328 EUR
APOSOR953395-1G APOSOR953395-250MG
4-Acetyl-1H-pyrrole-2-carboxylic acid 95%
4-Acetyl-1H-pyrrole-2-carboxylic acid
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571849
CAS-Nummer: 16168-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR