[2-(1-Methylpiperidin-4-yl)ethyl]amine dihydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
2-(1-Methylpiperidin-4-yl)ethanamine
OR953396-1G
OR953396-5G
APOSOR953396-1GEA
219
EUR
APOSOR953396-1G
APOSOR953396-5G
[2-(1-Methylpiperidin-4-yl)ethyl]amine dihydrochloride 95%
[2-(1-Methylpiperidin-4-yl)ethyl]amine dihydrochloride
Formel:
C₈H₂₀Cl₂N₂ Molecular Weight: 215,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09455608 CAS-Nummer: 20845-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...