3-(5-Methylpyrazol-1-yl)propionic acid 95%
Lieferant: Apollo Scientific
OR953399-1G
OR953399-250MG
APOSOR953399-1GEA
435
EUR
APOSOR953399-1G
APOSOR953399-250MG
3-(5-Methylpyrazol-1-yl)propionic acid 95%
3-(5-Methylpyrazol-1-yl)propionic acid
Formel:
C₇H₁₀N₂O₂ Molecular Weight: 154,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00996156 CAS-Nummer: 180741-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...