2',6'-Dihydroxy-4'-methylacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 3,5-Dihydroxy-4-acetyltoluene

OR953477-1G OR953477-5G
APOSOR953477-1GEA 92.8 EUR
APOSOR953477-1G APOSOR953477-5G
2',6'-Dihydroxy-4'-methylacetophenon 95%
2',6'-Dihydroxy-4'-methylacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.1739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656138
CAS-Nummer: 1634-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR