(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%

Lieferant: Apollo Scientific

Synonyme: Rasagiline, (1R)-N-prop-2-ynylindan-1-amine

OR953478-10G OR953478-1G OR953478-5G
APOSOR953478-10GEA 0 EUR
APOSOR953478-10G APOSOR953478-1G APOSOR953478-5G
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine
Formel: C₁₂H₁₃N
Molecular Weight: 171.2383 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00866571
CAS-Nummer: 136236-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR