1-tert-Butylazetidin-3-ol
Lieferant: Apollo Scientific
Synonyme:
1-tert-Butyl-3-azetidinol
OR953557-25G
OR953557-50G
OR953557-5G
APOSOR953557-25GEA
310
EUR
APOSOR953557-25G
APOSOR953557-50G
APOSOR953557-5G
1-tert-Butylazetidin-3-ol
1-tert-Butylazetidin-3-ol
Formel:
C₇H₁₅NO Molecular Weight: 129,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02323245 CAS-Nummer: 13156-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...