3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde ≥98%

Lieferant: Apollo Scientific

OR953566-100MG OR953566-1G OR953566-250MG
APOSOR953566-100MGEA 287 EUR
APOSOR953566-100MG APOSOR953566-1G APOSOR953566-250MG
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde ≥98%
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde
Formel: C₈H₅IN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08457789
CAS-Nummer: 900514-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR