(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine 95%
Lieferant: Apollo Scientific
OR953579-1G
OR953579-5G
APOSOR953579-1GEA
63.8
EUR
APOSOR953579-1G
APOSOR953579-5G
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine 95%
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine
Formel:
C₁₂H₁₈N₂ Molecular Weight: 190,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191310 CAS-Nummer: 169749-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...