Ethyl-1,2-benzoxazol-3-carboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
1,2-Benzisoxazole-3-carboxylic acid ethyl ester, 3-Ethoxycarbonyl-1,2-benzisoxazole, Ethyl 1,2-Benzisoxazole-3-carboxylate
OR953650-1G
OR953650-250MG
APOSOR953650-1GEA
719
EUR
APOSOR953650-1G
APOSOR953650-250MG
Ethyl-1,2-benzoxazol-3-carboxylat 95%
Ethyl benzo[d]isoxazole-3-carboxylate
Formel:
C₁₀H₉NO₃ Molecular Weight: 191.1834 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659787 CAS-Nummer: 57764-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...