1-Methyl-1H-indole-6-carbaldehyde 95%
Lieferant: Apollo Scientific
OR953712-1G
OR953712-250MG
APOSOR953712-1GEA
88.6
EUR
APOSOR953712-1G
APOSOR953712-250MG
1-Methyl-1H-indole-6-carbaldehyde 95%
1-Methyl-1H-indole-6-carbaldehyde
Formel:
C₁₀H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690251 CAS-Nummer: 21005-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...