2-Imidazo[1,2-a]pyridin-2-ylethanamine 95%

Lieferant: Apollo Scientific

OR953872-1G OR953872-250MG
APOSOR953872-1GEA 712 EUR
APOSOR953872-1G APOSOR953872-250MG
2-Imidazo[1,2-a]pyridin-2-ylethanamine 95%
2-Imidazo[1,2-a]pyridin-2-ylethanamine
Formel: C₉H₁₁N₃
Molecular Weight: 161.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738922
CAS-Nummer: 43170-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR