2-Methyl-5-oxo-1-cyclopentenyl butyrate 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-5-oxocyclopent-1-en-1-yl butyrate, Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester

OR953884-1G OR953884-5G
APOSOR953884-1GEA 52.2 EUR
APOSOR953884-1G APOSOR953884-5G
2-Methyl-5-oxo-1-cyclopentenyl butyrate 95%
2-Methyl-5-oxo-1-cyclopentenyl butyrate
Formel: C₁₀H₁₄O₃
Molecular Weight: 182,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798052
CAS-Nummer: 68227-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR