5-Chloro-2,3-dihydrobenzofuran-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR953943-1G OR953943-250MG
APOSOR953943-1GEA 632 EUR
APOSOR953943-1G APOSOR953943-250MG
5-Chloro-2,3-dihydrobenzofuran-2-carboxylic acid 95%
5-Chloro-2,3-dihydrobenzofuran-2-carboxylic acid
Formel: C₉H₇ClO₃
Molecular Weight: 198,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06753266
CAS-Nummer: 34385-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR