4-[(Methylamino)methyl]benzonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
4-((Methylamino)methyl)benzonitrile
OR953944-1G
OR953944-5G
APOSOR953944-1GEA
66.7
EUR
APOSOR953944-1G
APOSOR953944-5G
4-[(Methylamino)methyl]benzonitrile 98%
4-[(Methylamino)methyl]benzonitrile
Formel:
C₉H₁₀N₂ Molecular Weight: 146,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09738490 CAS-Nummer: 34403-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...