N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine 98%
Lieferant: Apollo Scientific
Synonyme:
1-(4-(Aminomethyl)phenyl)-N,N-dimethylmethanamine, 4-Dimethylaminomethylbenzylamine
OR953945-1G
APOSOR953945-1GEA
140
EUR
APOSOR953945-1G
N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine 98%
N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine
Formel:
C₁₀H₁₆N₂ Molecular Weight: 164.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04107485 CAS-Nummer: 34490-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...