Pyrrolidine-1-carbothioamide 98%

Lieferant: Apollo Scientific

OR954024-10G OR954024-1G OR954024-5G
APOSOR954024-10GEA 928 EUR
APOSOR954024-10G APOSOR954024-1G APOSOR954024-5G
Pyrrolidine-1-carbothioamide 98%
Pyrrolidine-1-carbothioamide
Formel: C₅H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00178436
CAS-Nummer: 40398-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR