(Indan-5-yloxy)acetic acid 96%

Lieferant: Apollo Scientific

Synonyme: 2-((2,3-Dihydro-1H-inden-5-yl)oxy)acetic acid

OR954050-10G OR954050-1G OR954050-5G
APOSOR954050-10GEA 1010 EUR
APOSOR954050-10G APOSOR954050-1G APOSOR954050-5G
(Indan-5-yloxy)acetic acid 96%
(Indan-5-yloxy)acetic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01672404
CAS-Nummer: 1878-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR