Methyl-2-pyridinylacetat 95%
Lieferant: Apollo Scientific
Synonyme:
2-Pyridineacetic Acid Methyl Ester, Methyl 2-Pyridineacetate, Methyl 2-(pyridin-2-yl)acetate, 2-Pyridylacetic Acid Methyl Ester
OR954071-25G
APOSOR954071-25GEA
98.6
EUR
APOSOR954071-25G
Methyl-2-pyridinylacetat 95%
Methyl-2-pyridinylacetat
Formel:
C₈H₉NO₂ Molecular Weight: 151,17 g/mol Siedepunkt: 137 °C Dichte: 1,13 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006358 CAS-Nummer: 1658-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...