3-(Benzyloxy)propan-1-amine 95%

Lieferant: Apollo Scientific

OR954152-1G OR954152-250MG
APOSOR954152-1GEA 328 EUR
APOSOR954152-1G APOSOR954152-250MG
3-(Benzyloxy)propan-1-amine 95%
3-(Benzyloxy)propan-1-amine
Formel: C₁₀H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07339550
CAS-Nummer: 16728-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR