Methyl-4-bromo-1H-indole-2-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-1H-indole-2-carboxylate

OR954166-1G OR954166-5G
APOSOR954166-1GEA 46.4 EUR
APOSOR954166-1G APOSOR954166-5G
Methyl-4-bromo-1H-indole-2-carboxylate 98%
Methyl-4-bromo-1H-indole-2-carboxylate
Formel: C₁₀H₈BrNO₂
Molecular Weight: 254.09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04966956
CAS-Nummer: 167479-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR