3-Nitrothioanisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(methylsulphanyl)-3-nitrobenzene

OR954517-25G OR954517-5G
APOSOR954517-25GEA 178 EUR
APOSOR954517-25G APOSOR954517-5G
3-Nitrothioanisol 95%
3-Nitrothioanisol
Formel: C₇H₇NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00024501
CAS-Nummer: 2524-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR