4-Phenylpyridin-2-ol 98%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxy-4-phenylpyridine
OR954648-1G
OR954648-250MG
APOSOR954648-1GEA
219
EUR
APOSOR954648-1G
APOSOR954648-250MG
4-Phenylpyridin-2-ol 98%
4-Phenylpyridin-2-ol
Formel:
C₁₁H₉NO Molecular Weight: 171,1953 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437954 CAS-Nummer: 19006-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...