1-(tert-Butyldimethylsilyl)indole-4-boronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-(tert-Butyldimethylsilyl)-1H-indol-4-yl)boronic acid

OR9553-1G OR9553-5G
APOSOR9553-1GEA 165 EUR
APOSOR9553-1G APOSOR9553-5G
1-(tert-Butyldimethylsilyl)indole-4-boronic acid 98%
1-(tert-Butyldimethylsilyl)indole-4-boronic acid
Formel: C₁₄H₂₂BNO₂Si
Molecular Weight: 275.23 g/mol
Schmelzpunkt: 130…135 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08436007
CAS-Nummer: 351457-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR