4-(Acetylsulphamoyl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9556-1G
OR9556-250MG
APOSOR9556-1GEA
973
EUR
APOSOR9556-1G
APOSOR9556-250MG
4-(Acetylsulphamoyl)benzeneboronic acid 98%
4-(Acetylsulphamoyl)benzeneboronic acid
Formel:
C₈H₁₀BNO₅S Molecular Weight: 243.04 g/mol Schmelzpunkt: 130…134 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436010 CAS-Nummer: 913835-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...