Methyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 96%

Lieferant: Apollo Scientific

OR955705-10G OR955705-1G OR955705-5G
APOSOR955705-10GEA 864 EUR
APOSOR955705-10G APOSOR955705-1G APOSOR955705-5G
Methyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 96%
Methyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Formel: C₈H₁₁NO₂
Molecular Weight: 153,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01553045
CAS-Nummer: 52459-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR