Methyl-2-amino-2-cyclopropylacetate hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-amino-2-cyclopropylacetate hydrochloride, Methyl 2-amino-2-cyclopropylacetate HCl

OR955907-1G OR955907-5G
APOSOR955907-1GEA 104 EUR
APOSOR955907-1G APOSOR955907-5G
Methyl-2-amino-2-cyclopropylacetate hydrochloride 95%
Methyl-2-amino-2-cyclopropylacetate hydrochloride
Formel: C₆H₁₂ClNO₂
Molecular Weight: 165,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031219
CAS-Nummer: 535936-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR