6-Acetamido-3-brompicolinsäure 96%
Lieferant: Apollo Scientific
Synonyme:
6-Acetamido-3-bromopyridine-2-carboxylic acid
OR9562-1G
OR9562-5G
APOSOR9562-1GEA
118
EUR
APOSOR9562-1G
APOSOR9562-5G
6-Acetamido-3-brompicolinsäure 96%
6-Acetamido-3-brompicolinsäure
Formel:
C₈H₇BrN₂O₃ Schmelzpunkt: 196…198 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235101 CAS-Nummer: 882430-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...