1-Nitro-4-propoxybenzol 98%
Lieferant: Apollo Scientific
Synonyme:
p-Nitrophenyl propyl ether
OR956283-1G
OR956283-5G
APOSOR956283-1GEA
219
EUR
APOSOR956283-1G
APOSOR956283-5G
1-Nitro-4-propoxybenzol 98%
1-Nitro-4-propoxybenzol
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
7244-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...