1-tert-Butyl-3-nitrobenzol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(tert-Butyl)-3-nitrobenzene

OR956369-5G
APOSOR956369-5GEA 63.8 EUR
APOSOR956369-5G
1-tert-Butyl-3-nitrobenzol 95%
1-tert-Butyl-3-nitrobenzol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086569
CAS-Nummer: 23132-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR