2-Chlor-4-methylanisol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-1-methoxy-4-methylbenzene, 3-Chloro-4-methoxytoluene

OR956946-1G OR956946-5G
APOSOR956946-1GEA 60.9 EUR
APOSOR956946-1G APOSOR956946-5G
2-Chlor-4-methylanisol 95%
2-Chlor-4-methylanisol
Formel: C₈H₉ClO
Molecular Weight: 156.609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060649
CAS-Nummer: 22002-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR