3-(4-Chlorphenyl)propiononitril 95%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Chlorophenyl)propanenitrile
OR957004-1G
OR957004-5G
APOSOR957004-1GEA
128
EUR
APOSOR957004-1G
APOSOR957004-5G
3-(4-Chlorphenyl)propiononitril 95%
3-(4-Chlorphenyl)propiononitril
Formel:
C₉H₈ClN Molecular Weight: 165,6195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310802 CAS-Nummer: 32327-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...