(4-Chlorphenoxy)essigsäure 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-CPA
OR957092-2.5KG
OR957092-500G
OR957092-100G
APOSOR957092-2.5KGEA
386
EUR
APOSOR957092-2.5KG
APOSOR957092-500G
APOSOR957092-100G
(4-Chlorphenoxy)essigsäure 98+%
(4-Chlorphenoxy)essigsäure
Formel:
(C₆H₄Cl)OCH₂COOH Molecular Weight: 186.59 g/mol Siedepunkt: 308 Schmelzpunkt: 156…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004305 CAS-Nummer: 122-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...