3,5-Dimethyl-1H-pyrazol-4-ol 95%

Lieferant: Apollo Scientific

OR957432-1G OR957432-5G
APOSOR957432-1GEA 111 EUR
APOSOR957432-1G APOSOR957432-5G
3,5-Dimethyl-1H-pyrazol-4-ol 95%
3,5-Dimethyl-1H-pyrazol-4-ol
Formel: C₅H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00030421
CAS-Nummer: 2820-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR