5-Methoxy-m-kresol 95%
Lieferant: Apollo Scientific
Synonyme:
3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzol, 3-Hydroxy-5-methoxytoluol, 3-Methoxy-5-methylphenol
OR957484-5G
APOSOR957484-5GEA
202
EUR
APOSOR957484-5G
5-Methoxy-m-kresol 95%
5-Methoxy-m-kresol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059261 CAS-Nummer: 3209-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...