Methyl-4-brompicolinat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-bromopicolinate, Methyl 4-bromopyridine-2-carboxylate
OR9577-25G
OR9577-1G
OR9577-5G
APOSOR9577-25GEA
149
EUR
APOSOR9577-25G
APOSOR9577-1G
APOSOR9577-5G
Methyl-4-brompicolinat 97%
Methyl-4-brompicolinat
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,03 g/mol Schmelzpunkt: 54…56 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07374897 CAS-Nummer: 29681-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...