3-Amino-5-brompyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Brom-3-pyridylamin, 5-Brompyridin-3-amin

OR9578-100G OR9578-25G OR9578-5G
APOSOR9578-100GEA 203 EUR
APOSOR9578-100G APOSOR9578-25G APOSOR9578-5G
3-Amino-5-brompyridin
3-Amino-5-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Schmelzpunkt: 65…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646060
CAS-Nummer: 13535-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR