4-Dimethylaminomethylbenzoic acid hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
4-((Dimethylamino)methyl)benzoic acid hydrochloride
OR958387-1G
OR958387-5G
APOSOR958387-1GEA
163
EUR
APOSOR958387-1G
APOSOR958387-5G
4-Dimethylaminomethylbenzoic acid hydrochloride 97%
4-Dimethylaminomethylbenzoic acid hydrochloride
Formel:
C₁₀H₁₄ClNO₂ Molecular Weight: 215.68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09997604 CAS-Nummer: 17847-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...