6-Phenoxypyridine-2-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Phenoxypicolinic acid

OR958421-1G OR958421-250MG OR958421-5G
APOSOR958421-1GEA 236 EUR
APOSOR958421-1G APOSOR958421-250MG APOSOR958421-5G
6-Phenoxypyridine-2-carboxylic acid 95+%
6-Phenoxypyridine-2-carboxylic acid
Formel: C₁₂H₉NO₃
Molecular Weight: 215,21 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14585023
CAS-Nummer: 51362-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR