2-Methyl-2-phenyloxiran 95%
Lieferant: Apollo Scientific
Synonyme: alpha-Methylstyrene Oxide, 1,2-Epoxy-2-phenylpropane
OR958718-1G OR958718-250MG
APOSOR958718-1GEA 72.5 EUR
APOSOR958718-1G APOSOR958718-250MG
2-Methyl-2-phenyloxiran 95%
2-Phenylpropylene oxide
Formel: C₉H₁₀O
Molecular Weight: 134.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059156
CAS-Nummer: 2085-88-3