2-Ethoxy-5-nitrobenzonitril 95%

Lieferant: Apollo Scientific

OR958805-1G OR958805-5G
APOSOR958805-1GEA 69.6 EUR
APOSOR958805-1G APOSOR958805-5G
2-Ethoxy-5-nitrobenzonitril 95%
2-Ethoxy-5-nitrobenzonitril
Formel: C₉H₈N₂O₃
Molecular Weight: 192,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02256084
CAS-Nummer: 29669-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR