N-(3-Hydroxypyridin-2-yl)acetamide 96%

Lieferant: Apollo Scientific

Synonyme: 2-Acetamido-3-hydroxypyridine

OR959383-5G
APOSOR959383-5GEA 75.4 EUR
APOSOR959383-5G
N-(3-Hydroxypyridin-2-yl)acetamide 96%
N-(3-Hydroxypyridin-2-yl)acetamide
Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09757484
CAS-Nummer: 31354-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR