5-Brom-2-cyclopropylpyrimidin 95%

Lieferant: Apollo Scientific

OR959384-1G OR959384-250MG
APOSOR959384-1GEA 134 EUR
APOSOR959384-1G APOSOR959384-250MG
5-Brom-2-cyclopropylpyrimidin 95%
5-Bromo-2-cyclopropylpyrimidine
Formel: C₇H₇BrN₂
Molecular Weight: 199,0479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702784
CAS-Nummer: 304902-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR