6-Chlor-3-formyl-2-picolin 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-methylpyridine-3-carbaldehyde, 6-Chloro-3-formyl-2-methylpyridine, 6-Chloro-2-methylnicotinaldehyde

OR959420-1G OR959420-5G
APOSOR959420-1GEA 30.6 EUR
APOSOR959420-1G APOSOR959420-5G
6-Chlor-3-formyl-2-picolin 98%
6-Chlor-3-formyl-2-picolin
Formel: C₇H₆ClNO
Molecular Weight: 155,5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368876
CAS-Nummer: 884495-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR