2-Chloro-5-methylnicotinaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-methylpyridine-3-carbaldehyde

OR959452-1G OR959452-250MG
APOSOR959452-1GEA 114 EUR
APOSOR959452-1G APOSOR959452-250MG
2-Chloro-5-methylnicotinaldehyde 98%
2-Chloro-5-methylnicotinaldehyde
Formel: C₇H₆ClNO
Molecular Weight: 155.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375370
CAS-Nummer: 92444-99-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR